Learn More
Starna Cells Inc SUB MICRO FLUORIMTR CELL 10UM
SDP
Catalog No. NC9990412
Encompass_Preferred
Sub-Micro Quartz Fluorometer Cell, 10 ul, 10mm Light Path, Z15
Catalog No. NC9990412 Supplier Starna Cells Inc Supplier No. 16.10FQ10/Z15
Sub-Micro Quartz Fluorometer Cell, 10 ul, 10mm Light Path, Z15