Learn More
Starna Cells Inc Sub-Micro Fluorometer Cells, 10 mm, 100 uL
SDP
Catalog No. NC1697539
Encompass_Preferred
Small sample chamber, 2mm wide, in fully masked standard exterior size cell with an emmision window 10mm wide. Fluorescent measurement using microliter volumes. Can be used with most fluorometers.
Catalog No. NC1697539 Supplier Starna Cells Inc Supplier No. 16.100FQ10/Z15
Small sample chamber, 2mm wide, in fully masked standard exterior size cell with an emmision window 10mm wide. Fluorescent measurement using microliter volumes. Can be used with most fluorometers.