Learn More
Starna Cells Inc Sub-Micro Fluorometer Cells, 40 uL
SDP
Catalog No. NC1697538
Encompass_Preferred
Small sample chamber, 2mm wide, in fully masked standard exterior size cell with an emmision window 10mm wide. Fluorescent measurement using microliter volumes.
Catalog No. NC1697538 Supplier Starna Cells Inc Supplier No. 16.40FQ10/Z15
Small sample chamber, 2mm wide, in fully masked standard exterior size cell with an emmision window 10mm wide. Fluorescent measurement using microliter volumes.