Learn More
Cole-Parmer Sub-Micro Quartz Cuvette, 100 uL, 8.5 mm Window Height; Each
Catalog No. NC1614865 Shop All Cole-Parmer Products
Encompass_Preferred
Available for UV/Vis absorbance spectroscopy applications (two polished windows). Match the cuvette center height to your instrument's light beam height; see individual cuvette specifications to verify correct manufacturer beam height.
Catalog No. NC1614865 Supplier Cole-Parmer Supplier No. 8330140
Available for UV/Vis absorbance spectroscopy applications (two polished windows). Match the cuvette center height to your instrument's light beam height; see individual cuvette specifications to verify correct manufacturer beam height.