Learn More
Hoefer, Inc SUB Swift Horizontal Electrophoresis System
SDP
Catalog No. NC3942890
Encompass_Preferred
The SUB Swift is a flexible, efficient, and user-friendly horizontal electrophoresis Unit.Supports workflows from single samples to 96-well high-throughput runs, both routine and large-scale applications. Features and Benefits:High Throughput – Run up to 108 samples; compatible with multichannel pipettesPlug-Free Lid – Open easily, protect electrodesVersatile Gel Casting UV-Transparent Tray Safe & Portable SpecificationsGel Dimensions (w x l) :13 x 13 cm, 13 x 6.5 cm, 6.5 x 6.5 cmBase Buffer: 300 mlSample Capacity : 108Maximum Power Settings : 300 V, 200 mA, 60 WUnit Dimensions (l x w x h) : 32 x 20 x 7 cmSafety Certifications.: CEIncludes:Gel tankSafety lidLarge gel tray, 13 x 13 cm, 1 pcs Medium gel tray, 13 x 6.5 cm, 1 pcsMini gel trays, 6.5 x 6.5 cm, 2 pcsLarge comb 12+12/27 wells, 1 mm thick, 4 pcsMedium comb 9+9/19 wells, 1.5 mm thick, 1 pcsMedium comb 7+7/14 wells, 1.5 mm thick, 1 pcsMini comb 2+3/3+3 wells, 2 mm thick, 1 pcs• Casting stand
Catalog No. NC3942890 Supplier Hoefer, Inc Supplier No. SUB SWIFT
The SUB Swift is a flexible, efficient, and user-friendly horizontal electrophoresis Unit.Supports workflows from single samples to 96-well high-throughput runs, both routine and large-scale applications. Features and Benefits:High Throughput – Run up to 108 samples; compatible with multichannel pipettesPlug-Free Lid – Open easily, protect electrodesVersatile Gel Casting UV-Transparent Tray Safe & Portable SpecificationsGel Dimensions (w x l) :13 x 13 cm, 13 x 6.5 cm, 6.5 x 6.5 cmBase Buffer: 300 mlSample Capacity : 108Maximum Power Settings : 300 V, 200 mA, 60 WUnit Dimensions (l x w x h) : 32 x 20 x 7 cmSafety Certifications.: CEIncludes:Gel tankSafety lidLarge gel tray, 13 x 13 cm, 1 pcs Medium gel tray, 13 x 6.5 cm, 1 pcsMini gel trays, 6.5 x 6.5 cm, 2 pcsLarge comb 12+12/27 wells, 1 mm thick, 4 pcsMedium comb 9+9/19 wells, 1.5 mm thick, 1 pcsMedium comb 7+7/14 wells, 1.5 mm thick, 1 pcsMini comb 2+3/3+3 wells, 2 mm thick, 1 pcs• Casting stand