Learn More
Andwin Scientific SUB ZERO BLUE FLIPPER RACK
SDP
Catalog No. NC9515347
Encompass_Preferred
The GeneMate IsoFreeze Flipper Rack provides secure cool storage for PCR reagents, cells, enzymes, or other temperature-sensitive reagents. This product is ideal for moving the items from the freezer to the benchtop or for events when the freezer experiences power failures. This rack can keep a temperature between -15C and -20C on the benchtop for up to 3 hours.
Catalog No. NC9515347 Supplier Andwin Scientific Supplier No. R20202
The GeneMate IsoFreeze Flipper Rack provides secure cool storage for PCR reagents, cells, enzymes, or other temperature-sensitive reagents. This product is ideal for moving the items from the freezer to the benchtop or for events when the freezer experiences power failures. This rack can keep a temperature between -15C and -20C on the benchtop for up to 3 hours.