Learn More
Sigma Aldrich Fine Chemicals Biosciences Suba-Seal(R) septa blue rubber, Suba Seal, 25, fits neck I.D., 14 mm, for use with 14/20 joints | 100EA
Catalog No. NC1830346
Encompass
Suba-Seal(R) septa blue rubber, Suba Seal, 25, fits neck I.D., 14 mm, for use with 14/20 joints | 100EA
Catalog No. NC1830346 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z648906100EA
Suba-Seal(R) septa blue rubber, Suba Seal, 25, fits neck I.D., 14 mm, for use with 14/20 joints | 100EA