Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #21.
SDP
Catalog No. 5090116581
Encompass
50pcs. Suba-Seal #21. . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 12.5mm Dimensions A 13.3mm B 9.6mm Strem# 96-1500. http://www.strem.com/catalog/v/96-1500/
Catalog No. 50-901-16581 Supplier Strem, An Ascensus Company Supplier No. 96150050PC
50pcs. Suba-Seal #21. . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 12.5mm Dimensions A 13.3mm B 9.6mm Strem# 96-1500. http://www.strem.com/catalog/v/96-1500/