Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #25 (fits female 14/20 joint).
SDP
Catalog No. 5090116587
Encompass
50pcs. Suba-Seal #25 (fits female 14/20 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 14.0mm Dimensions A 14.1mm B 12.5mm Strem# 96-1900. http://www.strem.com/catalog/v/96-1900/
Catalog No. 50-901-16587 Supplier Strem, An Ascensus Company Supplier No. 96190050PC
50pcs. Suba-Seal #25 (fits female 14/20 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 14.0mm Dimensions A 14.1mm B 12.5mm Strem# 96-1900. http://www.strem.com/catalog/v/96-1900/