Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #33 (fits Schlenk tube with male 24/40 joint).
SDP
Catalog No. 5090116589
Encompass
50pcs. Suba-Seal #33 (fits Schlenk tube with male 24/40 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 17.5mm Dimensions A 18.0mm B 12.7mm Strem# 96-3000. http://www.strem.com/catalog/v/96-3000/
Catalog No. 50-901-16589 Supplier Strem, An Ascensus Company Supplier No. 96300050PC
50pcs. Suba-Seal #33 (fits Schlenk tube with male 24/40 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 17.5mm Dimensions A 18.0mm B 12.7mm Strem# 96-3000. http://www.strem.com/catalog/v/96-3000/