Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #45 (fits female 24/40 joint).
SDP
Catalog No. 5090116596
Encompass
50pcs. Suba-Seal #45 (fits female 24/40 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 22.0mm Dimensions A 23.0mm B 20.4mm Strem# 96-3800. http://www.strem.com/catalog/v/96-3800/
Catalog No. 50-901-16596 Supplier Strem, An Ascensus Company Supplier No. 96380050PC
50pcs. Suba-Seal #45 (fits female 24/40 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 22.0mm Dimensions A 23.0mm B 20.4mm Strem# 96-3800. http://www.strem.com/catalog/v/96-3800/