Learn More
Sigma Aldrich Fine Chemicals Biosciences Suba SealR septa red rubbe
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z124664100EA
Encompass
Well-known as the best quality rubber turn-over closure with serrated stopper Suba-Seal brand is available exclusively from Sigma-Aldrich. Ideal for air and moisture-sensitive chemistry procedures. After piercing with non-coring needle rubber closes puncture providing air and moisture tight seal to protect contents of vessel or container from atmosphere.This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck making each serration a suctions sealing point.Storage Note for Natural Rubber Septa Store septa in a closed bag out of sunlight to retard blooming which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.Note for extra security when using large bore transfer needles fa
Catalog No. 50-290-4731