Learn More
Chemglass Life Sciences Septum Stopper, Suba-Seal, For 19/22-19/38 Outer Joints and 17.5mm ID Tubing
SDP
Catalog No. CG302403
Encompass_Preferred
Septum Stopper, Suba-Seal, For 19/22-19/38 Outer Joints and 17.5mm ID Tubing. Suba-Seal septa are a flanged stopper with a hollow plug and sleeve extension that folds down over the neck of the vessel. In addition to the pull over sleeve, Suba-Seal stoppers have a series of annular serrations on the plug portion of the septa. Each of these serrations depress along the inner wall of the glass vessel to form an extremely effective air-tight seal. Stoppers are made from natural red rubber and sold in the package quantities listed below.
This item is not returnable. View return policy
Catalog No. CG302403 Supplier Chemglass Life Sciences Supplier No. CG302403
Septum Stopper, Suba-Seal, For 19/22-19/38 Outer Joints and 17.5mm ID Tubing. Suba-Seal septa are a flanged stopper with a hollow plug and sleeve extension that folds down over the neck of the vessel. In addition to the pull over sleeve, Suba-Seal stoppers have a series of annular serrations on the plug portion of the septa. Each of these serrations depress along the inner wall of the glass vessel to form an extremely effective air-tight seal. Stoppers are made from natural red rubber and sold in the package quantities listed below.