Learn More
Medchemexpress LLC HY-111789 10mg , Subasumstat CAS:1858276-04-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11178910MG
Encompass_Preferred
HY-111789 10mg Subasumstat CAS: 1858276-04-6 Subasumstat (TAK-981) is a first in class and selective inhibitor of the SUMOylation enzymatic cascade, with potential immune-activating and antineoplastic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9311