Learn More
Medchemexpress LLC Subasumstat | 1858276-04-6 | 578.10 | 100 MG
SDP
Catalog No. 5000437342
Encompass_Preferred
Subasumstat (TAK-981) is a first-in-class and selective inhibitor of the SUMOylation enzymatic cascade, demonstrating potential immune-activating and antineoplastic activities. It increases the production of type 1 IFN, enhancing type 1 IFN-mediated signaling, activating innate effector cells, and improving antitumor innate immune responses. A single subcutaneous injection induces activation of dendritic cells.
  • First-in-class selective inhibitor of SUMOylation enzymatic cascade
  • Potential immune-activating activities
  • Potential antineoplastic activities
  • Increases production of type 1 IFN
  • Enhances antitumor innate immune responses
  • Activates innate effector cells
  • Induces activation of dendritic cells
Catalog No. 50-004-37342 Supplier Medchemexpress LLC Supplier No. HY111789100MG
Add to Cart
Add to Cart
Subasumstat (TAK-981) is a first-in-class and selective inhibitor of the SUMOylation enzymatic cascade, demonstrating potential immune-activating and antineoplastic activities. It increases the production of type 1 IFN, enhancing type 1 IFN-mediated signaling, activating innate effector cells, and improving antitumor innate immune responses. A single subcutaneous injection induces activation of dendritic cells.
  • First-in-class selective inhibitor of SUMOylation enzymatic cascade
  • Potential immune-activating activities
  • Potential antineoplastic activities
  • Increases production of type 1 IFN
  • Enhances antitumor innate immune responses
  • Activates innate effector cells
  • Induces activation of dendritic cells