Learn More
Grainger Subbase Metal/Plastic 265/208/230V
Catalog No. 502764237
Encompass_Preferred
The Grainger 36PT88 is a 3/4 HP, 115V, 1725 RPM, 56 frame, open drip proof, ball bearing, C-face motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a NEMA standard C-face mounting with a standard shaft. The motor is designed for general purpose applications.
Catalog No. 50-276-4237 Supplier Grainger Supplier No. 36PT88
The Grainger 36PT88 is a 3/4 HP, 115V, 1725 RPM, 56 frame, open drip proof, ball bearing, C-face motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a NEMA standard C-face mounting with a standard shaft. The motor is designed for general purpose applications.