Learn More
Novus Biologicals™ Subcellular Protein Fractionation Kit

Description
Specifications
Specifications
| Includes | Protein Fractionation Kit |
| Product Type | Subcellular Fractions |
| Target Proteins | Subcellular Protein Fractionation |
| Content And Storage | Store at -20°C Avoid freeze-thaw cycles |
| For Use With (Application) | 1-D or 2-D gel, enzyme activity assays, gel supershift assay, and Western blotting; Subcellular Protein Fractionation: The Cell Fractionation Kit can be used in 1-D or 2-D gel, enzyme activity assays, gel shift assay, and Western blotting. |
| Quantity | 50 Assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.