Learn More
Zen Bio Inc Subcutaneous Adipocytes 96-Well Plate, BMI >30.0
SDP
Catalog No. NC1582503
Encompass
ZenBio provides the highest quality primary human cultured preadipocytes and adipocytes. Our cells have multiple applications in basic research, drug screening and development. Cells are isolated from a wide variety of patients with good representation from all levels of adiposity and age. Cells are available from both male and female donors. Pooled lots of cells are available from both male and female donors to provide large lots for screening and to decrease issues with patient to patient variability. Preadipocytes are provided cryopreserved or in culture. Adipocytes are differentiated in vitro from preadipocytes using our patent protected differentiation methods. Cells are available in a variety of plate formats. All cells have passed our rigorous quality control measures including performance in lipid accumulation, lipolysis, and adipokine secretion. Cells have tested negative for HIV-1, HIV-2, HTLV-1, HTLV-2, Hepatitis B, Hepatitis C and mycoplasma.
Catalog No. NC1582503 Supplier Zen Bio Inc Supplier No. SA10963
ZenBio provides the highest quality primary human cultured preadipocytes and adipocytes. Our cells have multiple applications in basic research, drug screening and development. Cells are isolated from a wide variety of patients with good representation from all levels of adiposity and age. Cells are available from both male and female donors. Pooled lots of cells are available from both male and female donors to provide large lots for screening and to decrease issues with patient to patient variability. Preadipocytes are provided cryopreserved or in culture. Adipocytes are differentiated in vitro from preadipocytes using our patent protected differentiation methods. Cells are available in a variety of plate formats. All cells have passed our rigorous quality control measures including performance in lipid accumulation, lipolysis, and adipokine secretion. Cells have tested negative for HIV-1, HIV-2, HTLV-1, HTLV-2, Hepatitis B, Hepatitis C and mycoplasma.