Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Suberic acid

Catalog No. S5200100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S5200100G 100 g
S5200500G 500 g
S52005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S5200100G Supplier Sigma Aldrich Supplier No. S5200100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Suberic acid

Specifications

CAS 505-48-6
Melting Point 139°C to 146°C, 140°C to 144°C (lit.)
Boiling Point 230°C (15 mmHg)
Molecular Formula C8H14O4
Linear Formula HOOC(CH2)6 COOH
MDL Number MFCD00004428
Quantity 100 g
Synonym Octanedioic acid
Molecular Weight (g/mol) 174.19
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.