Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Suberic Acid Bis(3-Sulfo-N-Hydroxysuccinimide Ester) Sodium Salt

Catalog No. S5799100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S5799100MG 100 mg
S579925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S5799100MG Supplier Sigma Aldrich Supplier No. S5799100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is a water soluble, homobifunctional cross-linking reagent with amine reactivity. It is typically coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates 8-ato mLinker.Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is useful for the preparation of protein-protein, receptor-ligand and hapten carrier molecule conjugates.

Specifications

CAS 82436-77-9
Molecular Formula C16H18N2Na2O14S2
Linear Formula C16H18N2Na2O14S2
Quantity 100 mg
MDL Number MFCD00054982
Synonym BS3; Bis(sulfosuccinimidyl) suberate sodium salt
Molecular Weight (g/mol) 572.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.