Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Suberic Acid Bis(N-Hydroxysuccinimide Ester)

Catalog No. S18855G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S18855G 1 g
S1885500MG 5 g
S18851G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S18855G Supplier Sigma Aldrich Supplier No. S18855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Suberic acid bis(N-hydroxysuccinimide ester) is a homobifunctional cross-linking reagent with amine reactivity. Typically coupled to molecules containing primary amine by amide bond buffered at pH 7.5 (6.5-8.5). Suberic acid bis(N-hydroxysuccinimide ester) incorporates an 8-ato mLinker.Suberic acid bis(N-hydroxysuccinimide ester) has been:supplemented in pierce lysis buffer used for immunoprecipitationused to conjugate the oligonucleotide standards to gelatinused in cross-linking of ASC (apoptosis-associated speck-like protein containing a caspase recruitment domain) oligomers

Specifications

CAS 68528-80-3
Molecular Formula C16H20N2O8
Linear Formula C16H20N2O8
MDL Number MFCD00049059
Quantity 1 g
Synonym Disuccinimidyl suberate; Disuccinimidyl octanedioate; Disuccinimidyl suberate (DSS)
Molecular Weight (g/mol) 368.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.