Learn More
Medchemexpress LLC Suberic acid-d12 | 169397-99-3 | 98.6% | 186.27 | C8H2D12O4 | 50 MG
SDP
Catalog No. 5000303259
Encompass_Preferred
Suberic acid-d12 is the deuterium-labeled form of suberic (octanedioic) acid intended for use as a stable isotope internal standard and tracer in mass spectrometry and analytical workflows. It is supplied as a white to off-white solid with high isotopic and chemical purity, suitable for quantitative analyses, method development, and metabolomics research.
  • Deuterium-labeled analogue for accurate internal standardization.
  • High chemical purity (98.6%) for reliable analytical performance.
  • White to off-white solid, easy to handle and store.
  • Suitable for mass spectrometry, tracer studies, and quantitative workflows.
  • Available in small research-scale quantities for method development.
Catalog No. 50-003-03259 Supplier Medchemexpress LLC Supplier No. HYW015300S150MG
Suberic acid-d12 is the deuterium-labeled form of suberic (octanedioic) acid intended for use as a stable isotope internal standard and tracer in mass spectrometry and analytical workflows. It is supplied as a white to off-white solid with high isotopic and chemical purity, suitable for quantitative analyses, method development, and metabolomics research.
  • Deuterium-labeled analogue for accurate internal standardization.
  • High chemical purity (98.6%) for reliable analytical performance.
  • White to off-white solid, easy to handle and store.
  • Suitable for mass spectrometry, tracer studies, and quantitative workflows.
  • Available in small research-scale quantities for method development.