Learn More
Medchemexpress LLC Suberic acid-d4 | 19031-57-3 | 99.9% | 178.22 | C8H10D4O4 | 10 MG
SDP
Catalog No. 5000303734
Encompass_Preferred
Suberic acid-d4 is the deuterium-labeled analogue of suberic (octanedioic) acid used as an internal standard and reference material in mass spectrometry and metabolic studies. Supplied as an analytical-grade solid in small mg-scale packaging, it is intended for quantitative assays, method development, and metabolite identification.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic and chemical purity for accurate quantitation.
  • Suitable for quantitative assays and method development.
  • Stable solid form for convenient storage and handling.
  • Soluble in common solvents for sample preparation workflows.
Catalog No. 50-003-03734 Supplier Medchemexpress LLC Supplier No. HYW015300S10MG
Suberic acid-d4 is the deuterium-labeled analogue of suberic (octanedioic) acid used as an internal standard and reference material in mass spectrometry and metabolic studies. Supplied as an analytical-grade solid in small mg-scale packaging, it is intended for quantitative assays, method development, and metabolite identification.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic and chemical purity for accurate quantitation.
  • Suitable for quantitative assays and method development.
  • Stable solid form for convenient storage and handling.
  • Soluble in common solvents for sample preparation workflows.