Learn More
Medchemexpress LLC Suberic acid-d4 (octanedioic acid-d4) | 19031-57-3 | 99.9% | 178.22 | C8H10D4O4 | 50 MG
SDP
Catalog No. 5000303735
Encompass_Preferred
Suberic acid-d4 (octanedioic acid-d4) is the deuterated analogue of suberic acid supplied as a high-purity solid for use as a stable isotope internal standard in analytical workflows. It is intended for LC-MS quantification, metabolic tracing, and analytical method development.
  • Deuterium-labeled stable isotope standard.
  • High chemical and isotopic purity (99.93%).
  • Solid form with white to light yellow appearance.
  • Suitable for LC-MS and method development applications.
  • Available in small analytical package sizes, including 50 mg.
  • Recommended storage: powder at -20°C (stable up to 3 years).
Catalog No. 50-003-03735 Supplier Medchemexpress LLC Supplier No. HYW015300S50MG
Add to Cart
Add to Cart
Suberic acid-d4 (octanedioic acid-d4) is the deuterated analogue of suberic acid supplied as a high-purity solid for use as a stable isotope internal standard in analytical workflows. It is intended for LC-MS quantification, metabolic tracing, and analytical method development.
  • Deuterium-labeled stable isotope standard.
  • High chemical and isotopic purity (99.93%).
  • Solid form with white to light yellow appearance.
  • Suitable for LC-MS and method development applications.
  • Available in small analytical package sizes, including 50 mg.
  • Recommended storage: powder at -20°C (stable up to 3 years).