Learn More
Medchemexpress LLC Suberic acid (octanedioic acid) | 505-48-6 | MFCD00004428 | 98.0% | 174.20 | C8H14O4 | 100 G

Supplier: Medchemexpress LLC HYW015300100G
Suberic acid (octanedioic acid) is a crystalline aliphatic dicarboxylic acid (CAS 505-48-6) used as a reagent and building block in organic synthesis and biochemical research. Supplied as a solid with analytical-grade purity, it is suitable for synthetic chemistry, formulation studies, and laboratory assays.
- High purity (98.0%).
- Solid, white to off-white appearance.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Storage stability specified for powder and in-solution conditions.
- Available in common laboratory pack sizes, including 100 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.