Learn More
Medchemexpress LLC Suberic acid (octanedioic acid) | 505-48-6 | MFCD00004428 | 98.0% | 174.20 g/mol | C8H14O4 | 50 G
SDP
Catalog No. 5000302524
Encompass_Preferred
Suberic acid (octanedioic acid) is a saturated dicarboxylic acid provided as a crystalline solid for research use. It is used as an analytical reference and in biochemical studies, reported to modulate Akt signaling and TGF-β receptor pathways. Key chemical identifiers include CAS 505-48-6, formula C8H14O4, molecular weight 174.20 g/mol, and typical purity of 98.0%.
  • Crystalline dibasic acid suitable for biochemical research.
  • Typical purity 98.0% for analytical applications.
  • Soluble in DMSO; offered as solid and solution formats.
  • Used as an endogenous metabolite reference and pathway probe.
  • Available in multiple pack sizes, including 50 g.
Catalog No. 50-003-02524 Supplier Medchemexpress LLC Supplier No. HYW01530050G
Add to Cart
Add to Cart
Suberic acid (octanedioic acid) is a saturated dicarboxylic acid provided as a crystalline solid for research use. It is used as an analytical reference and in biochemical studies, reported to modulate Akt signaling and TGF-β receptor pathways. Key chemical identifiers include CAS 505-48-6, formula C8H14O4, molecular weight 174.20 g/mol, and typical purity of 98.0%.
  • Crystalline dibasic acid suitable for biochemical research.
  • Typical purity 98.0% for analytical applications.
  • Soluble in DMSO; offered as solid and solution formats.
  • Used as an endogenous metabolite reference and pathway probe.
  • Available in multiple pack sizes, including 50 g.