Learn More
Cayman Chemical Suberohydroxamic acid
SDP
Catalog No. 502887239
Encompass_Preferred
A competitive HDAC inhibitor that inhibits HDAC1 (IC50 = 0.25 μM) and HDAC3 (IC50 = 0.30 μM); causes cell differentiation, cell cycle arrest, or apoptosis
Catalog No. 50-288-7239 Supplier Cayman Chemical Supplier No. 10574250 MG
Add to Cart
Add to Cart
A competitive HDAC inhibitor that inhibits HDAC1 (IC50 = 0.25 μM) and HDAC3 (IC50 = 0.30 μM); causes cell differentiation, cell cycle arrest, or apoptosis