Learn More
Enzo Life Sciences Suberoyl bis-hydroxamic acid (100mg). CAS: 38937-66-5
SDP
Catalog No. 502010974
Encompass_Preferred
Induces cellular differentiation. Inhibits histone deacetylation. Purity: ≥95% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0974 Supplier Enzo Life Sciences Supplier No. BMLGR3230100
Induces cellular differentiation. Inhibits histone deacetylation. Purity: ≥95% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.