Learn More
Medchemexpress LLC HY-W009776 50mg Medchemexpress, Suberoyl bis-hydroxamic acid CAS:38937-66-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW00977650MG
Encompass_Preferred
Medchemexpress, HY-W009776 50mg Suberoyl bis-hydroxamic acid CAS:38937-66-5 Suberoyl bis-hydroxamic acid (Suberohydroxamic acid; SBHA) is a competitive and cell-permeable HDAC1 and HDAC3 inhibitor with ID50 values of 0.25 μM and 0.30 μM, respectively.Suberoyl bis-hydroxamic acid renders MM cells susceptible to apoptosis and facilitates the mitochondrial apoptotic pathways.Suberoyl bis-hydroxamic acid can be used for the study of medullary thyroid carcinoma (MTC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3275