Learn More
Medchemexpress LLC Suberoyl bis-hydroxamic acid | 38937-66-5 | 95.0% | 204.22 | 250 MG
SDP
Catalog No. 5000453412
Encompass_Preferred
Suberoyl bis-hydroxamic acid is a competitive and cell-permeable HDAC1 and HDAC3 inhibitor. It renders MM cells susceptible to apoptosis, facilitating mitochondrial apoptotic pathways. This product can be used for the study of medullary thyroid carcinoma (MTC).
  • Competitive and cell-permeable HDAC1 and HDAC3 inhibitor
  • Renders MM cells susceptible to apoptosis
  • Facilitates mitochondrial apoptotic pathways
  • Suitable for the study of medullary thyroid carcinoma (MTC)
  • Appears as a white to off-white solid
Catalog No. 50-004-53412 Supplier Medchemexpress LLC Supplier No. 250MGHYW009776
Suberoyl bis-hydroxamic acid is a competitive and cell-permeable HDAC1 and HDAC3 inhibitor. It renders MM cells susceptible to apoptosis, facilitating mitochondrial apoptotic pathways. This product can be used for the study of medullary thyroid carcinoma (MTC).
  • Competitive and cell-permeable HDAC1 and HDAC3 inhibitor
  • Renders MM cells susceptible to apoptosis
  • Facilitates mitochondrial apoptotic pathways
  • Suitable for the study of medullary thyroid carcinoma (MTC)
  • Appears as a white to off-white solid