Learn More
Medchemexpress LLC Suberoylanilide hydroxamic acid | 149647-78-9 | MFCD00945317 | 99.9% | 264.32 | C14H20N2O3 | 250 MG
SDP
Catalog No. 5000247158
Encompass_Preferred
Vorinostat (suberoylanilide hydroxamic acid, SAHA) analytical standard supplied as a 250 mg solid reference material for research and analytical applications. It is intended for assay development, calibration, and analytical method validation for HDAC inhibitor studies.
  • Analytical standard for method development and calibration.
  • Approximately 99.9% purity for reliable reference measurements.
  • Solid, white to off-white appearance for straightforward handling.
  • Molecular formula C14H20N2O3 and molecular weight 264.32 included for identification.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-47158 Supplier Medchemexpress LLC Supplier No. HY10221R250MG
Vorinostat (suberoylanilide hydroxamic acid, SAHA) analytical standard supplied as a 250 mg solid reference material for research and analytical applications. It is intended for assay development, calibration, and analytical method validation for HDAC inhibitor studies.
  • Analytical standard for method development and calibration.
  • Approximately 99.9% purity for reliable reference measurements.
  • Solid, white to off-white appearance for straightforward handling.
  • Molecular formula C14H20N2O3 and molecular weight 264.32 included for identification.
  • Intended for research use only; not for human or clinical use.