Learn More
Cayman Chemical Suberylglycine, Purity: ≥95%, CAS: 60317-54-6, 100 mg
SDP
Catalog No. 502948167
Encompass_Preferred
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency
Catalog No. 50-294-8167 Supplier Cayman Chemical Supplier No. 34619100 MG
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency