Learn More
Cayman Chemical SuberylglycIn 10mg
SDP
Catalog No. 502950961
Encompass_Preferred
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency
Catalog No. 50-295-0961 Supplier Cayman Chemical Supplier No. 3461910 MG
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency