Learn More
Cayman Chemical Suberylglycine, Purity: ≥95%, CAS: 60317-54-6, 50 mg
SDP
Catalog No. 502950266
Encompass_Preferred
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency
Catalog No. 50-295-0266 Supplier Cayman Chemical Supplier No. 3461950 MG
A glycine-conjugated form of suberic acid; urinary levels are increased in patients with MCAD deficiency