Learn More
Medchemexpress LLC Suberylglycine-d4 | 2469259-73-0 | 99.0% | 235.27 | 1 MG
SDP
Catalog No. 5000437052
Encompass_Preferred
Suberylglycine-d4 is a deuterium-labeled form of Suberylglycine, which is an acyl glycine and a minor metabolite of fatty acid. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-37052 Supplier Medchemexpress LLC Supplier No. HY113367S1MG
Suberylglycine-d4 is a deuterium-labeled form of Suberylglycine, which is an acyl glycine and a minor metabolite of fatty acid. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.