Learn More
Chemglass Life Sciences Sublimation Adapter Complete, 24/25
SDP
Catalog No. CG303201
Encompass_Preferred
Sublimation Adapter Complete, 24/25. Adapter permits sublimation directly in any reaction vessel having an standard taper outer joint; simply remove condenser or other apparatus from joint, insert adapter and begin. Adapter has two components, the cold finger condenser and vacuum adapter. Adapter has a #18 Chem-Thread to allow for the vertical positioning of the cold finger in flasks from 250 to 1000mL. Vacuum is controlled via a 2mm PTFE stopcock. Adapter joint sizes are medium length, but can be used in full length joints.
Catalog No. CG303201 Supplier Chemglass Life Sciences Supplier No. CG303201
Sublimation Adapter Complete, 24/25. Adapter permits sublimation directly in any reaction vessel having an standard taper outer joint; simply remove condenser or other apparatus from joint, insert adapter and begin. Adapter has two components, the cold finger condenser and vacuum adapter. Adapter has a #18 Chem-Thread to allow for the vertical positioning of the cold finger in flasks from 250 to 1000mL. Vacuum is controlled via a 2mm PTFE stopcock. Adapter joint sizes are medium length, but can be used in full length joints.