Learn More
Chemglass Life Sciences Sublimator Complete, Minum-Ware
SDP
Catalog No. 501532696
Encompass_Preferred
Sublimator Complete, Minum-Ware. Sublimator consisting of a MW-33 adapter and vertically adjustable condenser tube having an O.D. of 7mm. For use with all 14/10 jointed flasks and reaction vials. Supplied complete with nylon compression cap (CG-352-01) and Viton o-ring (CG-305-07).
Catalog No. 50-153-2696 Supplier Chemglass Life Sciences Supplier No. MW7301
Sublimator Complete, Minum-Ware. Sublimator consisting of a MW-33 adapter and vertically adjustable condenser tube having an O.D. of 7mm. For use with all 14/10 jointed flasks and reaction vials. Supplied complete with nylon compression cap (CG-352-01) and Viton o-ring (CG-305-07).