Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Flask, Round Bottom, Long Neck 25 mL
GREENER_CHOICESDP

Catalog No. 31502088 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-088 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-088 Supplier Synthware™ Supplier No. DF302425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Borosilicate glass flask

Special 25 mL long neck flask for small volume efficiency.

Specifications

Color Clear
Capacity (Metric) 25 mL
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Includes Long neck round bottom flask
Product Type Boiling Flask
Standard Taper Joint 24/40
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Round
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.