Learn More
Synthware™ Sublimination Apparatus
GREENER_CHOICESDP
Catalog No. p-7230073 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
100 mL
150 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-085 100 mL
31-502-086 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-085 Supplier Synthware™ Supplier No. S740100
Only null left
Add to Cart
Add to Cart
Two-piece grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer
Condenser has a large top opening for use with dry ice/acetone or liquid nitrogen. Connection between the flask and condenser is made with a flat O-ring joint permitting parts to be gently separated for maximum product recovery. The capacity listed is that below the bottom of the cold finger. Vacuum control is provided by a 0 to 8 mm high vacuum valve. Complete apparatus consists of the condenser, flask, Viton™ O-ring and horseshoe clamp.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Quantity 1 Ea.
Capacity (Metric) 100 mL
Product Type Dry Ice/Acetone/Liquid Nitrogen