Learn More
Hach Company Submersion Mounting Hardware
Supplier: Hach Company MH434B
CPVC pipe (1 inch diameter x 1 2m (4ft) long) and coupling with PVC pipe mount junction box for use with LCP pH/ORP sensors