Learn More
Grainger Submersion pH Kit Ryton PPS BNC Flat Tip
Catalog No. 502768169
Encompass_Preferred
The SENSOREX S8100 10 BNC Submersion pH Sensor is a pH electrode designed for laboratory and industrial applications. It has a glass pH sensor, double junction Ag/AgCl reference, and BNC connector. The sensor has a measurement range of 0-14 pH, accuracy of ±0.02 pH, and temperature range of 0-80°C. It has a built-in temperature sensor for automatic temperature compensation. The electrode body is made of epoxy and is waterproof. It has a cable length of 1 meter. The S8100 can be fully submerged and is ideal for measurements in liquids. It requires a pH meter for operation.
Catalog No. 50-276-8169 Supplier Grainger Supplier No. 16Z031
The SENSOREX S8100 10 BNC Submersion pH Sensor is a pH electrode designed for laboratory and industrial applications. It has a glass pH sensor, double junction Ag/AgCl reference, and BNC connector. The sensor has a measurement range of 0-14 pH, accuracy of ±0.02 pH, and temperature range of 0-80°C. It has a built-in temperature sensor for automatic temperature compensation. The electrode body is made of epoxy and is waterproof. It has a cable length of 1 meter. The S8100 can be fully submerged and is ideal for measurements in liquids. It requires a pH meter for operation.