Learn More
Polysciences, Inc. Submicron Bead Calibration Kit (0.2µm, 0.5µm, 0.8µm)
SDP
Catalog No. NC3717672
Encompass
Current applications in flow cytometry extend far beyond traditional lymphocyte immunophenotyping, with some involving the analysis of very small particles such as platelet- and endothelial-derived microparticles or microbial species. Our Small Bead Calibration Kits allow operators to verify the resolution capabilities of the flow cytometer, and to establish appropriate instrument settings for the analyses. For more Flow Cytometry Resources, visit the Bangs Flow Blog.Nominal Diameters: 0.2μm, 0.5μm, 0.8μmParticle Concentration: 1 x 108 particles/mlFluorescence: YG (488nm excitation, green emission)
Catalog No. NC3717672 Supplier Polysciences, Inc. Supplier No. BLI8321
Current applications in flow cytometry extend far beyond traditional lymphocyte immunophenotyping, with some involving the analysis of very small particles such as platelet- and endothelial-derived microparticles or microbial species. Our Small Bead Calibration Kits allow operators to verify the resolution capabilities of the flow cytometer, and to establish appropriate instrument settings for the analyses. For more Flow Cytometry Resources, visit the Bangs Flow Blog.Nominal Diameters: 0.2μm, 0.5μm, 0.8μmParticle Concentration: 1 x 108 particles/mlFluorescence: YG (488nm excitation, green emission)