Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences MXS Calibration Standard 301BE for AFM, SEM Mounted, 300 nm Spacing
SDP

Catalog No. 5029158
Encompass_Preferred

EMS offers a series of calibration standards with one and two dimension calibrated patterns. These standards are created utilizing holographic interference of a particular laser frequency. They are typically accurate to <1% across the entire surface of the standard. These reference standards are remarkably durable under typical operating conditions. The surface contamination behavior is also very good. There are no better sub-micron reference standard available in this price range. For AFM, SEM, TOF-SIMS, Auger
Mount: Mounted
Spacing: 300 nm
Structure: Parallel Ridges
Material: Ti lines on silica

Catalog No. 50-291-58 Supplier Electron Microscopy Sciences Supplier No. 8011131M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EMS offers a series of calibration standards with one and two dimension calibrated patterns. These standards are created utilizing holographic interference of a particular laser frequency. They are typically accurate to <1% across the entire surface of the standard. These reference standards are remarkably durable under typical operating conditions. The surface contamination behavior is also very good. There are no better sub-micron reference standard available in this price range. For AFM, SEM, TOF-SIMS, Auger
Mount: Mounted
Spacing: 300 nm
Structure: Parallel Ridges
Material: Ti lines on silica

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.