Learn More
Medchemexpress LLC Substance P(1-7) TFA | 2828433-22-1 | 99.1% | 1014.06 | 25 MG
SDP
Catalog No. 5000426792
Encompass_Preferred
Substance P(1-7) TFA is a fragment of the neuropeptide, substance P (SP). It gives depressor and bradycardic effects when applied to the nucleus tractus solitarius. Substance P(1-7) is found to act as a very potent antagonist against the SP-induced responses and is formed locally in the nigra after SP injection. It is proposed that Substance P(1-7) is an endogenous modulator of SP actions. For research use only.
  • Fragment of neuropeptide, Substance P (SP)
  • Acts as a potent antagonist against SP-induced responses
  • Exhibits depressor and bradycardic effects
  • Modulates actions of SP or SP(5-11) in the spinal cord
  • For research use only
Catalog No. 50-004-26792 Supplier Medchemexpress LLC Supplier No. 25MGHYP1485A
Substance P(1-7) TFA is a fragment of the neuropeptide, substance P (SP). It gives depressor and bradycardic effects when applied to the nucleus tractus solitarius. Substance P(1-7) is found to act as a very potent antagonist against the SP-induced responses and is formed locally in the nigra after SP injection. It is proposed that Substance P(1-7) is an endogenous modulator of SP actions. For research use only.
  • Fragment of neuropeptide, Substance P (SP)
  • Acts as a potent antagonist against SP-induced responses
  • Exhibits depressor and bradycardic effects
  • Modulates actions of SP or SP(5-11) in the spinal cord
  • For research use only