Learn More
Medchemexpress LLC HY-P1485A 5mg Medchemexpress, Substance P (1-7)(TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1485A5MG
Encompass_Preferred
Medchemexpress, HY-P1485A 5mg Substance P (1-7)(TFA) CAS: Substance P (1-7)(TFA) is a fragment of the neuropeptide, substance P (SP). Substance P (1-7)(TFA) gives depressor and bradycardic effects when applied to the nucleus tractus solitarius. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4212