Learn More
Eurofins Discoverx Substance P, 1 mg. (92-1069)
Supplier:  Eurofins Discoverx 921069
Encompass
Substance P is an 11 amino acid sensory tachykinin neuropeptide present in the CNS that acts as a neurotransmitter and neuromodulator of inflammation and pain. Substance P is an agonist for TACR1, 2 and 3. Activation involves binding to neurokinin-1 receptor (NK-1R). Plays a role in the constriction of bronchioles. Provided as 1 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9609