Learn More
Medchemexpress LLC HY-P0201 1mg Medchemexpress, Substance P CAS:33507-63-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02011MG
Encompass_Preferred
Medchemexpress, HY-P0201 1mg Substance P CAS:33507-63-0 Substance P (Neurokinin P) is a neuropeptide, acting as a neurotransmitter and as a neuromodulator in the CNS. The endogenous receptor for substance P is neurokinin 1 receptor (NK1-receptor, NK1R). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3028