Learn More
CPC Scientific H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2 (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific SUBS001C
Encompass
This neuropeptide was found in the gut as well as in the brain. It is responsible for a number of excitatory effects on both central and peripheral neurons. It contracts smooth muscle, constricts bronchioles and increases capillary permeability. When released from afferent nerves, it causes a neurogenic inflammatory response, including mast cell degranulation.SEQUENCE: H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: RPKPQQFFGLM-NH2MOLECULAR FORMULA: C63H98N18O13S1MOLECULAR WEIGHT: 1347.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [33507-63-0]SYNONYMS: RESEARCH AREA: Neuropeptides, Pain, Inflammation, HormonalREFERENCES:A.I. Roberts et al., Cell Immunol., 141, 457 (1992)SKU(s): SUBS-001A, SUBS-001B, SUBS-001C
Catalog No. 50-211-0457