Learn More
Medchemexpress LLC Substance P | 33507-63-0 | >99.6% | 1347.63 | 5 MG
SDP
Catalog No. 5000425888
Encompass_Preferred
Substance P (Neurokinin P) is a CNS-penetrant neuropeptide that functions as both a neurotransmitter and a neuromodulator in the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R).
  • CNS-penetrant neuropeptide
  • Acts as a neurotransmitter and neuromodulator in the CNS
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Available in multiple package sizes
  • High purity of 99.6%
Catalog No. 50-004-25888 Supplier Medchemexpress LLC Supplier No. HYP02015MG
Substance P (Neurokinin P) is a CNS-penetrant neuropeptide that functions as both a neurotransmitter and a neuromodulator in the central nervous system. Its endogenous receptor is the neurokinin 1 receptor (NK1-receptor, NK1R).
  • CNS-penetrant neuropeptide
  • Acts as a neurotransmitter and neuromodulator in the CNS
  • Endogenous receptor is neurokinin 1 receptor (NK1R)
  • Available in multiple package sizes
  • High purity of 99.6%