Learn More
Enzo Life Sciences Substance P (5'-TAMRA conjugate) (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLSX58000001
Encompass_Preferred
Fluorescent photostable ligand for binding to the neurokinin-1 receptor. Useful for localizing the NK1R in live tissue slice preparations. Alternative name: TMR-SP, Detection probe for neurokinin 1 receptor. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO (at least 10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2203